missing translation for 'onlineSavingsMsg'
Learn More

Adipic dihydrazide, 97%

Catalog No. AAA1511914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1511914 25 g
AAA1511922 100 g
AAA1511936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1511914 Supplier Thermo Scientific Chemicals Supplier No. A1511914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1071-93-8 | C6H14N4O2 | 174.20 g/mol

Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adipic dihydrazide is used as a formaldehyde scavenger and reacts with formaldehyde, thereby preventing the volatilizing of formaldehyde in the air. It is also employed as a paint additive and coating additive. It is also used as an intermediate. Further, it is used for cross-linking water-based emulsions and as a hardener for certain epoxy resins, which finds application in powder coating.

Solubility
Soluble in water, alcohol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN

Specifications

Melting Point 180°C to 182°C
Odor Odorless
Quantity 25 g
Beilstein 973863
Solubility Information Soluble in water,alcohol and acetone.
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material Adipic dihydrazide
Hazard Category H317-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P272-P280g-P302+P352-P304+P340-P312-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 213-999-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only