missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid 99.0+%, TCI America™

Catalog No. p-7116746
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016125G 25 g
A0161500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A016125G Supplier TCI America Supplier No. A016125G
Only null left

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00004420
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
Synonym adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german
PubChem CID 196
ChEBI CHEBI:30832
IUPAC Name hexanedioic acid
SMILES C(CCC(=O)O)CC(=O)O

Specifications

pH 3.2 (0.1% H2O soln./25°C)
Melting Point 153°C
Color White
Formula Weight 146.14
Percent Purity ≥99.0% (GC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Adipic Acid
EINECSNumber (2)-0858
RTECSNumber AU8400000
TSCA Yes