missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 153°C |
| Color | White |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 146.14 |
| Percent Purity | ≥99.0% (GC,T) |
| Chemical Name or Material | Adipic Acid |
Safety and Handling
| EINECSNumber | (2)-0858 |
| RTECSNumber | AU8400000 |
| TSCA | Yes |