missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid, Honeywell Fluka™
Meets analytical specification of Ph.Eur., BP, E 355, 99.6-101.0% (calc. to the dried substance)
$783.13 - $783.13
Chemical Identifiers
| CAS | 124-04-9 |
|---|---|
| Molecular Formula | C6H10O4 |
| Molecular Weight (g/mol) | 146.142 |
| MDL Number | MFCD00004420 |
| InChI Key | WNLRTRBMVRJNCN-UHFFFAOYSA-N |
| Synonym | adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german |
| PubChem CID | 196 |
| ChEBI | CHEBI:30832 |
| IUPAC Name | hexanedioic acid |
| SMILES | C(CCC(=O)O)CC(=O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 124-04-9 | |
| 146.142 | |
| WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
| 196 | |
| hexanedioic acid |
| C6H10O4 | |
| MFCD00004420 | |
| adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
| CHEBI:30832 | |
| C(CCC(=O)O)CC(=O)O |
Specifications
| 124-04-9 | |
| HOOC(CH2)4COOH | |
| 1209788 | |
| Freely soluble in methanol, boiling water, cyclohexane, acetone, water, ethanol. Insoluble in benzene,petroleum ether. | |
| C(CCC(=O)O)CC(=O)O | |
| 146.142 | |
| CHEBI:30832 | |
| 99.6 to 101% (calc. to the dried substance) | |
| 196°C (384.8°F) | |
| ≤0.002% (as SO4) | |
| 1mmHg (159.5°C) |
| C6H10O4 | |
| MFCD00004420 | |
| adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
| WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
| hexanedioic acid | |
| 196 | |
| 146.14g/mol | |
| 25 kg | |
| 788°F | |
| 5 (vs air) | |
| Adipic Acid, Meets analytical specification of Ph.Eur.BP |
Safety and Handling
P305 + P351 + P338
H319
EINECSNumber : 204-673-3