missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid, FCC, 99.6-101%, Spectrum™ Chemical

Catalog No. p-7156993
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600321 25 g Poly Bottle
18600325 100 g Poly Bottle
18600322 500 g Poly Bottle
18600323 3 kg Poly Pail
18600326 12 kg Poly Pail
18600324 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600321 Supplier Spectrum Chemical Mfg Cor Supplier No. AD13025GM
Only null left

AD130, 124-04-9, C6H10O4

Spectrum™ Chemical Adipic Acid, FCC is used as a food ingredient as a flavorant and gelling aid and as an acidulant in baking powders. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 124-04-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
InChI Key WNLRTRBMVRJNCN-UHFFFAOYSA-N
IUPAC Name hexanedioic acid
SMILES OC(=O)CCCCC(O)=O

Specifications

CAS Max % 100%
Percent Purity 99.6 to 101%
Grade FCC
Quantity 25 g
Ignition Residue 0.002%
Packaging Poly Bottle