missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adipic Acid, FCC, 99.6-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Adipic Acid, FCC is used as a food ingredient as a flavorant and gelling aid and as an acidulant in baking powders. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 45 kg |
| Ignition Residue | 0.002% |
| Packaging | Fiber Drum |
| Percent Purity | 99.6 to 101% |
| Grade | FCC |