Learn More
Adipic acid, 99%
CAS: 124-04-9 | C6H10O4 | 146.142 g/mol
Supplier: Thermo Scientific Chemicals 102810010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Adipic acid | |
| 124-04-9 | |
| 98.5 | |
| White | |
| C6H10O4 | |
| MFCD00004420 | |
| 01,15 | |
| adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
| C(CCC(=O)O)CC(=O)O | |
| 146.142 | |
| CHEBI:30832 | |
| 99% | |
| 196°C | |
| Plastic bottle |
| 151.0°C to 153.0°C | |
| 337.0°C | |
| 100.0 | |
| 99% | |
| HO2C(CH2)4CO2H | |
| 02, 649 | |
| 15, 150 | |
| WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
| hexanedioic acid | |
| 196 | |
| 146.14 | |
| 1 kg | |
| Authentic | |
| Crystalline Powder |
Chemical Identifiers
| 124-04-9 | |
| 146.142 | |
| WNLRTRBMVRJNCN-UHFFFAOYSA-N | |
| 196 | |
| hexanedioic acid |
| C6H10O4 | |
| MFCD00004420 | |
| adipic acid, adipinic acid, 1,4-butanedicarboxylic acid, adilactetten, acifloctin, acinetten, 1,6-hexanedioic acid, molten adipic acid, kyselina adipova, adipinsaure german | |
| CHEBI:30832 | |
| C(CCC(=O)O)CC(=O)O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only