missing translation for 'onlineSavingsMsg'
Learn More

Adipamide 98.0+%, TCI America™

Catalog No. A016025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A016025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A016025G Supplier TCI America Supplier No. A016025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-94-4
Molecular Formula C6H12N2O2
Molecular Weight (g/mol) 144.174
MDL Number MFCD00008045
InChI Key GVNWZKBFMFUVNX-UHFFFAOYSA-N
Synonym adipamide, adipic diamide, adipic acid amide, adipic acid diamide, 1,4-butanedicarboxamide, 1,2-ethylenebisacetamide, unii-40dru033ow, ccris 5883, butane-1,4-dicarboxamide, hexanedioic diamide
PubChem CID 12364
IUPAC Name hexanediamide
SMILES C(CCC(=O)N)CC(=O)N

Specifications

Melting Point 229°C
Color White
Quantity 25 g
Formula Weight 144.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Adipamide
EINECSNumber (2)-0881
RTECSNumber AU7800000
TSCA Yes