missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adhesive Seals

Catalog No. AB2558
Change view
Click to view available options
Material:
Polyethylene
For Use With (Application):
Crystallography and Genotyping
Fluorescence Applications, QPCR
Sample storage, Thermal Cycling w/ Lid
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application)
AB2558 Polyethylene Sample storage, Thermal Cycling w/ Lid
AB2170 Polyethylene Fluorescence Applications, QPCR
AB2450 Polyethylene Crystallography and Genotyping
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AB2558 Supplier Thermo Scientific™ Supplier No. AB2558

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform high throughput applications at a lower cost per plate with these adhesive seals, featuring an automated format provides efficient sealing.

Perform high throughput applications at a lower cost per plate with Thermo Scientific™ Adhesive Seals. An automated format provides efficient sealing. Rolls are designed for use with the Thermo Scientific™ SEAL-IT 100™ adhesive sealer.

Adhesive PCR Film Roll
  • Clear adhesive seal prevents evaporation during sample storage or thermal cycling, while still allowing samples to be accessed by piercing foil with pipette tips
  • No adhesive build-up on tips after piercing
  • PCR Film seals are recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to 120°C

Adhesive PCR Foil Roll

  • Strong adhesive seal prevents evaporation during sample storage or thermal cycling, while allowing sample access by piercing foil with pipet tips
  • No adhesive build-up on tips after piercing
  • PCR Foil Seals are recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to +150°C

Adhesive PCR Film

  • Optically clear adhesive plate seal for sample storage; thermal cycling with screw down or clip down lid
  • Strong adhesive, peelable with no sticky residue left on the plate
  • Temperature range: -20°C to +120°C

ABsolute QPCR Seal Roll

  • Optically clear adhesive plate seal is designed for fluorescence applications, including QPCR
  • Features strong pressure-sensitive, non-sticky adhesive that prevents evaporation during sample storage or thermal cycling
  • Seals are easily peelable, but leave no sticky residue on the plate when removed
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to +110°C

Adhesive Crystallography Seal Roll

  • Optically clear, pressure sensitive, non-sticky adhesive seal for crystallography and genotyping applications
  • Samples are not exposed to adhesive
  • Seal is easily peelable without leaving residue on the plate
  • Temperature range: -70° to +110C°

Specifications

Color Clear
Description Adhesive PCR Film, Roll 80mmx100m
Material Polyethylene
For Use With (Application) Sample storage, Thermal Cycling w/ Lid
For Use With (Equipment) The Seal-it 100 adhesive sealer.
Product Type Adhesive PCR Film
Pierceable Yes
Temperature Range (Metric) -20°F to +120°C
Plate Compatibility 96/384-well Plate