Learn More
3M™ Adflo™ Spark Arrestor, 37142, 15-0009-99X02
Catalog No. 296365140
Change view
Click to view available options
2/Cs.
Catalog No.
296365140 2/Cs.
1 options
Catalog No. 296365140 Supplier 3M™ Supplier No. 7000126730
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement spark arrestor for 3M™ Adflo™ Powered Air Purifying Respirator.
  • Installs into the 3M™ Adflo™ Turbo Filter Cover.
  • Replacement part
Specifications
Product Type Spark Arrestors
Quantity 2/Cs.