missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-Triphosphate Disodium Salt Hydrate 98.0+%, TCI America™
Supplier: TCI America A015725G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 34369-07-8 | |
| 551.15 | |
| TTWYZDPBDWHJOR-WCYUCLFNNA-L | |
| 131664345 | |
| [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| C10H14N5Na2O13P3 | |
| MFCD00150755 | |
| atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal | |
| disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
Specifications
| Adenosine 5′-Triphosphate Disodium Salt Hydrate | |
| White | |
| MFCD00150755 | |
| atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal | |
| [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 551.15 | |
| 551.14 | |
| Crystalline Powder |
| 34369-07-8 | |
| C10H14N5Na2O13P3 | |
| 25 g | |
| TTWYZDPBDWHJOR-WCYUCLFNNA-L | |
| disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate | |
| 131664345 | |
| ≥98.0% (HPLC,T) |
Safety and Handling
EINECSNumber : (9)-0048
RTECSNumber : AU7417000
TSCA : Yes
Recommended Storage : Refrigerator