missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals
Adenosine-5'-triphosphate Disodium Salt Hydrate Cell culture reagent, ≥96%, White Powder
$93.89 - $2201.69
Chemical Identifiers
| CAS | 51963-61-2 |
|---|---|
| Molecular Formula | C10H20N5Na2O16P3 |
| Molecular Weight (g/mol) | 605.19 |
| MDL Number | MFCD00150754 |
| InChI Key | MWEQTWJABOLLOS-MVHBFMHHNA-L |
| PubChem CID | 134129625 |
| SMILES | O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15026601
|
MP Biomedicals Inc
0215026601 |
1 g |
Each for $93.89
|
|
|||||
|
ICN15026680
|
MP Biomedicals Inc
0215026680 |
100 g |
Each for $2,201.69
|
|
|||||
Description
- Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
Chemical Identifiers
| 51963-61-2 | |
| 605.19 | |
| MWEQTWJABOLLOS-MVHBFMHHNA-L | |
| O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| C10H20N5Na2O16P3 | |
| MFCD00150754 | |
| 134129625 |
Specifications
| White | |
| Powder | |
| C10H20N5Na2O16P3 | |
| adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt | |
| O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 605.19 | |
| 551.2 (Anhydrous) |
| 51963-61-2 | |
| 1 g | |
| MFCD00150754 | |
| MWEQTWJABOLLOS-MVHBFMHHNA-L | |
| disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate | |
| 134129625 | |
| Adenosine-5'-Triphosphate Disodium Salt Hydrate, Hydrate |
Safety and Handling
Recommended Storage : Store at 0°C.