missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals
Supplier: MP Biomedicals Inc 0219461401
Specifications
| Adenosine-5'-Triphosphate Disodium Salt Hydrate | |
| White | |
| Powder | |
| C10H20N5Na2O16P3 | |
| adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt | |
| O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
| 605.19 | |
| 551.2 (Anhydrous) |
| Hydrate | |
| 51963-61-2 | |
| 1 g | |
| MFCD00150754 | |
| MWEQTWJABOLLOS-MVHBFMHHNA-L | |
| disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate | |
| 134129625 |
Chemical Identifiers
| 51963-61-2 | |
| 605.19 | |
| MWEQTWJABOLLOS-MVHBFMHHNA-L | |
| 134129625 | |
| O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
| C10H20N5Na2O16P3 | |
| MFCD00150754 | |
| adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt | |
| disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate |
Safety and Handling
Recommended Storage : Store at 0°C.