missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98%
Description
Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.
Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.
Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 10 g |
| Sensitivity | Hygroscopic; Light sensitive |
| Merck Index | 14,155 |
| Solubility Information | Soluble in water (110mg/ml at 25°C),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),DMF (sparingly),and PBS pH 7.2 (∽10mg/ml). |
| Formula Weight | 551.15 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Adenosine-5'-triphosphate disodium salt hydrate |
Safety and Handling
| EINECSNumber | 213-579-1 |
| TSCA | Yes |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only