missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, 98%

Catalog No. AAJ6112506
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6112506 5 g
AAJ6112509 10 g
AAJ6112514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6112506 Supplier Thermo Scientific Chemicals Supplier No. J6112506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol

Adenosine-5'-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adenosine-5′-triphosphate disodium salt hydrate, is used as a P2 purinoceptor agonist.

Solubility
Soluble in water (110 mg/ml at 25°C), ethanol (<1mg/mL at 25°C), DMSO (<1mg/mL at 25°C), DMF (sparingly), and PBS pH 7.2 (∼10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 34369-07-8
Molecular Formula C10H14N5Na2O13P3
Molecular Weight (g/mol) 551.15
MDL Number MFCD00150755
InChI Key TTWYZDPBDWHJOR-WCYUCLFNNA-L
Synonym atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal
PubChem CID 131664345
IUPAC Name disodium;[[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-hydroxyphosphoryl] phosphate;hydrate
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,155
Solubility Information Soluble in water (110mg/ml at 25°C),ethanol (<1mg/ml at 25°C),DMSO (<1mg/ml at 25°C),DMF (sparingly),and PBS pH 7.2 (∽10mg/ml).
Formula Weight 551.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Adenosine-5'-triphosphate disodium salt hydrate
EINECSNumber 213-579-1
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only