missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-triphosphate Disodium Salt, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor AD1255KGBL

 View more versions of this product

Catalog No. 18608450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

987-65-5
Poly Pail
0.99
1
5 kg
Reagent

Chemical Identifiers

987-65-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.