New England Biolab Ltd Adenosine-5 Triphosphate (ATP) 1 ml
Catalog No. 50994931
Adenosine 5-Triphosphate (ATP) is nucleotide used as a substrate for many ATP-dependent enzyme systems, including protein kinases, protein phosphatases and DNA ligation.
Catalog No.
50994931
Supplier
New England Biolab LtdSupplier No.
P0756S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Adenosine 5-Triphosphate (ATP) is nucleotide used as a substrate for many ATP-dependent enzyme systems, including protein kinases, protein phosphatases and DNA ligation.