missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-monophosphoric acid, 99% (dry wt.), water <6%

CAS: 61-19-8 | C10H14N5O7P | 347.22 g/mol

Supplier:  Thermo Scientific Chemicals L1405106

 View more versions of this product

Catalog No. AAL1405106

Only null left
Explore available promotions

Description

Description

a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists. 5?-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5?-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
a useful ligand determinant that facilitate the binding of APS reductase inhibitors and activates adenosine receptor agonists. 5′-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase, an activator of phosphorylase b in the polymerization reaction for the synthesis of branched polysaccharides, a reagent in the synthesis of adenosine-5′-phosphoimidazolide, an inhibitor of endogenous NMN adenylyltransferase (NMNAT) activity that converts NMN to NAD+.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Specifications

Adenosine-5'-monophosphoric acid
∽190°C (decomposition)
(dry wt.), water <6%
MFCD00005750
5205540
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
UDMBCSSLTHHNCD-YPLCUDRINA-N
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
347.22
CHEBI:16027
99%
61-19-8
Odorless
C10H14N5O7P
5 g
14,158
Soluble in water.
−47° (c=2 in 2% NaOH)
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
6083
347.22

Chemical Identifiers

61-19-8
347.22
UDMBCSSLTHHNCD-YPLCUDRINA-N
6083
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
C10H14N5O7P
MFCD00005750
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
CHEBI:16027
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-500-0

RTECSNumber : AU7480500

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only