missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-monophosphate sodium salt hydrate, 98+%

Catalog No. AC440710250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC440710250 25 g Glass bottle
AC440710010 1 g Glass bottle
AC440710050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC440710250 Supplier Thermo Scientific Chemicals Supplier No. 440710250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149022-20-8 | C10H13N5NaO7P | 369.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149022-20-8
Molecular Formula C10H13N5NaO7P
Molecular Weight (g/mol) 369.21
MDL Number MFCD18785650
InChI Key FYWLYWMEGHCZAX-GWKNMROSNA-M
Synonym sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x, adenosine 5-monophosphoric acid disodium salt, adenosine 5'-monophosphate sodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, 5'-adenylic acid,sodium salt, hydrate 9ci, disodium hydrate adenosine-5-monophosphate 2-
PubChem CID 23674992
IUPAC Name sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
CAS Min % 98.5
pH 4.50 to 6.50
CAS Max % 100.0
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 369.21
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Adenosine 5′-monophosphate sodium salt hydrate
Show More Show Less

RUO – Research Use Only