missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-Diphosphate Trisodium Salt, MP Biomedicals
Supplier: MP Biomedicals Inc 0210004910
Specifications
| Adenosine-5'-Diphosphate Trisodium Salt | |
| C10H15N5Na3O10P2 | |
| adenosine-5'-diphosphate trisodium salt | |
| C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na] | |
| 496.172 | |
| 493.2 |
| 2092-65-1 | |
| 10 g | |
| VJZLETZZUNQROW-MSQVLRTGSA-N | |
| [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium | |
| 131875859 |
Chemical Identifiers
| 2092-65-1 | |
| 496.172 | |
| adenosine-5'-diphosphate trisodium salt | |
| [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate;sodium |
| C10H15N5Na3O10P2 | |
| VJZLETZZUNQROW-MSQVLRTGSA-N | |
| 131875859 | |
| C1=NC2=C(C(=N1)N)N=CN2C3C(C(C(O3)COP(=O)(O)OP(=O)(O)O)O)O.[Na].[Na].[Na] |