missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-diphosphate trilithium salt, 98%

CAS: 31008-64-7 | C10H12Li3N5O10P2 | 445.00 g/mol

Supplier:  Thermo Scientific Chemicals J6449706

Quantity 5 g

 View more versions of this product

Catalog No. AAJ6449706


Only null left

Description

Description

It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a salt formulation of the essential metabolic compound adenosine diphosphate (ADP).

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

31008-64-7
445.00
LZGPPAHUZSOGHJ-DJXXCMMGNA-K
56841973
[Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
C10H12Li3N5O10P2
MFCD00065469
adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
trilithium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate

Specifications

Adenosine-5'-diphosphate trilithium salt
White
5 g
adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
LZGPPAHUZSOGHJ-DJXXCMMGNA-K
trilithium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
56841973
98%
31008-64-7
C10H12Li3N5O10P2
MFCD00065469
Soluble in water.
[Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
445.00
445
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 250-431-5

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions