missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-diphosphate trilithium salt, 96 to 99%, For HPLC analysis, MP Biomedicals™

Catalog No. p-4404648
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15026101 1 g
ICN15026105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15026101 Supplier MP Biomedicals Inc Supplier No. 0215026101

Chemical Identifiers

CAS 31008-64-7
Molecular Formula C10H12Li3N5O10P2
Molecular Weight (g/mol) 445.00
MDL Number MFCD00065469
InChI Key LZGPPAHUZSOGHJ-DJXXCMMGNA-K
Synonym adenosine-5'-diphosphate trilithium salt, adenosine 5'-trihydrogen diphosphate , trilithium salt, adp-li3, trilithium 1+ adenosine 5'-diphosphate, adenosine 5'-diphosphate trilithium salt, adenosine 5'-diphosphate,trilithium salt, adenosine-5'-diphosphate, trilithium salt, trilithium 1+ ion adenosine 5'-diphosphate, adenosine 5'-diphosphoric acid trilithium salt, adenosine-5'-diphosphate,trilithium salt
PubChem CID 56841973
IUPAC Name trilithium(1+) ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Li+].[Li+].[Li+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 96 to 99%
Quantity 1 g
Formula Weight 454
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphosphate trilithium salt, For HPLC analysis