missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-diphosphate Monosodium Salt, 97.7%, MP Biomedicals™

Catalog No. p-4405466
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1901495 500 mg
ICN19014901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN1901495 Supplier MP Biomedicals Inc Supplier No. 02190149.5
Only null left

Chemical Identifiers

CAS 20398-34-9
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.20
MDL Number MFCD00066473,MFCD00065470,MFCD00066635
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adp monosodium salt, adenosine 5'-diphosphate, sodium salt, adenosine 5'-diphosphate sodium salt
PubChem CID 87704518
IUPAC Name [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 0.977
Quantity 500 mg
Formula Weight 449.19
Physical Form Powder
Chemical Name or Material Adenosine-5'-diphosphate Monosodium Salt