missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-5'-diphosphate monopotassium salt dihydrate, 99%

Catalog No. AAJ6067203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6067203 1 g
AAJ6067206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6067203 Supplier Thermo Scientific Chemicals Supplier No. J6067203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72696-48-1 | C10H14KN5O10P2 | 465.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
IUPAC Name potassium;(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolan-3-olate;dihydrate
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Quantity 1 g
Formula Weight 501.33 (465.29 Anhydrous)
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphosphate monopotassium salt dihydrate
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only