missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate Disodium Salt Dihydrate

Catalog No. AAJ1049006
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1049006 5 g Plastic bottle
AAJ1049014 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1049006 Supplier Thermo Fisher Scientific Supplier No. J1049006

CAS: 16178-48-6 | C10H13N5Na2O10P2 | 471.17 g/mol

Chemical Identifiers

CAS 16178-48-6
Molecular Formula C10H13N5Na2O10P2
Molecular Weight (g/mol) 471.17
MDL Number MFCD00150927
InChI Key ORKSTPSQHZNDSC-WCYUCLFNNA-L
Synonym adenosine 5 '-diphosphate disodium salt, adenosine-5 '-diphosphate disodium salt, sodium 2 r,3 s,4 r,5 r-5-6-amino-9 h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl hydrogendiphosphate, adenosine 5 '-diphosphate disodium salt hydrate, adenosine, 5 '-diphosphoric acid, disodium salt, adenosine 5'-trihydrogen diphosphate , disodium salt, trinosin s hydrate, disodium hydrogen 2r,3s,4r,5r-5-6-aminopurin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxyphosphonate, adenosine 5'-trihydrogen diphosphate , sodium salt 1:2, disodium adp hydrate
PubChem CID 85315
SMILES [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Physical Form Powder
Quantity 5 g
Beilstein 3645656
IUPAC Name disodium;[[(2 R,3 S,4 R,5 R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
Formula Weight 507.2
Packaging Plastic bottle
Chemical Name or Material Adenosine-5 '-Diphosphate Disodium Salt Dihydrate
EINECSNumber 240-314-7
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only