missing translation for 'onlineSavingsMsg'
Learn More

Adenine 99.0+%, TCI America™

Catalog No. a0149250g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
A0149250G 250 g
A014925G 25 g
2 options

Catalog No. A0149250G

Supplier: TCI America A0149250G

Only null left

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Melting Point 365°C
Color White-Yellow
Quantity 250 g
UN Number 2811
Formula Weight 135.13
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Adenine
EINECSNumber (9)-0045
RTECSNumber AU6125000
TSCA Yes