missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hydrochloride, Cell Culture Reagent Grade, MP Biomedicals™

Catalog No. ICN19460805
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19460805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19460805 Supplier MP Biomedicals Inc Supplier No. 0219460805

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6055-72-7
Molecular Formula C5H8ClN5O
Molecular Weight (g/mol) 189.60
MDL Number MFCD11046306
InChI Key MYRDTAUFFBYTHA-UHFFFAOYSA-N
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
PubChem CID 24211900
IUPAC Name 7H-purin-6-amine hydrate hydrochloride
SMILES O.Cl.NC1=C2NC=NC2=NC=N1

Specifications

Color White
Quantity 5 g
Formula Weight 171.6
Grade Cell Culture Reagent
Physical Form Solid
Chemical Name or Material Adenine Hydrochloride
Recommended Storage Store at room temperature.