missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hydrochloride, ∼99%, MP Biomedicals™

Catalog No. p-4403629
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10019105 5 g
ICN10019180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10019105 Supplier MP Biomedicals Inc Supplier No. 0210019105
Only null left

Adenine Hydrochloride, ∼99% is used in viral research and has a wide range of chemical and biochemical roles in vivo and in vitro.

  • Adenine has a wide range of chemical and biochemical roles in vivo and in vitro.
  • It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).
  • It is used as a blood preservative and in viral research.
  • It is a purine nucleobase widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids.
  • It is a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.

Chemical Identifiers

CAS 6055-72-7
Molecular Formula C5H8ClN5O
Molecular Weight (g/mol) 189.60
MDL Number MFCD11046306
InChI Key MYRDTAUFFBYTHA-UHFFFAOYSA-N
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
PubChem CID 24211900
IUPAC Name 7H-purin-6-amine hydrate hydrochloride
SMILES O.Cl.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance λ max 262 nm (0.1 N HCl)
Color White
Assay Percent Range 0.992
Quantity 5 g
Formula Weight 171.6
Percent Purity 99%
Physical Form Powder
For Use With (Application) Used as a blood preservative and in viral research, Regulatory molecule
Chemical Name or Material Adenine hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.