Learn More
Thermo Scientific Chemicals Adenine hydrochloride, 98+%, cont. up to ca 5% water
CAS: 2922-28-3 | C5H6ClN5 | 171.59 g/mol
Supplier: Thermo Scientific Chemicals A1762222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Adenine hydrochloride | |
| ∼290°C (decomposition) | |
| C5H6ClN5 | |
| 100 g | |
| adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1 | |
| Cl.NC1=C2NC=NC2=NC=N1 | |
| 171.59 | |
| 171.59 |
| 2922-28-3 | |
| cont. up to ≈5% water | |
| MFCD00038990 | |
| 4009585 | |
| UQVDQSWZQXDUJB-UHFFFAOYSA-N | |
| 7H-purin-6-amine;hydrochloride | |
| 76219 | |
| ≥98% |
Chemical Identifiers
| 2922-28-3 | |
| 171.59 | |
| UQVDQSWZQXDUJB-UHFFFAOYSA-N | |
| 76219 | |
| Cl.NC1=C2NC=NC2=NC=N1 |
| C5H6ClN5 | |
| MFCD00038990 | |
| adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1 | |
| 7H-purin-6-amine;hydrochloride |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 220-867-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only