missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine hydrochloride, 98+%, cont. up to ca 5% water

Catalog No. AAA1762206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1762206 5 g
AAA1762214 25 g
AAA1762222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1762206 Supplier Thermo Scientific Chemicals Supplier No. A1762206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2922-28-3 | C5H6ClN5 | 171.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2922-28-3
Molecular Formula C5H6ClN5
Molecular Weight (g/mol) 171.59
MDL Number MFCD00038990
InChI Key UQVDQSWZQXDUJB-UHFFFAOYSA-N
Synonym adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
PubChem CID 76219
IUPAC Name 7H-purin-6-amine;hydrochloride
SMILES Cl.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∼290°C (decomposition)
Quantity 5 g
Beilstein 4009585
Formula Weight 171.59
Percent Purity ≥98%
Assay cont. up to ≈5% water
Chemical Name or Material Adenine hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-867-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only