missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine, 99%

CAS: 73-24-5 | C5H5N5 | 135.13 g/mol

Supplier:  Thermo Scientific Chemicals A1490630

 View more versions of this product

Catalog No. AAA1490630

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Adenine is used as an active component of boron-deficient media to grow yeast in order to assess whether yeast growth is stimulated by boron. It is useful as a local antiseptic and vitamin B4. Further, it is used in the microbial determination of niacin. It is also employed as a food supplement for adult rats to investigate the effects of dietary adenine overload. In addition to this, it is used in the production of nucleotides of the nucleic acids.

Solubility
Soluble in hot water, methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

Adenine
>360°C (decomposition)
Odorless
MFCD00041790
UN2811
14,152
Soluble in hot water,methanol and ethanol. Slightly soluble in cold water. Insoluble in diethyl ether.
NC1=C2NC=NC2=NC=N1
135.13
CHEBI:16708
99%
73-24-5
99%
C5H5N5
250 g
5777
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
GFFGJBXGBJISGV-UHFFFAOYSA-N
7H-purin-6-amine
190
135.13

Chemical Identifiers

73-24-5
135.13
GFFGJBXGBJISGV-UHFFFAOYSA-N
190
7H-purin-6-amine
C5H5N5
MFCD00041790
adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
CHEBI:16708
NC1=C2NC=NC2=NC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-796-1

RTECSNumber : AU6125000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only