Learn More
Thermo Scientific Chemicals Adenine, 99%
CAS: 73-24-5 | C5H5N5 | 135.13 g/mol
Supplier: Thermo Scientific Chemicals 147440250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Adenine | |
| 99.5 | |
| 100.0 | |
| Colorless to Yellow | |
| Authentic | |
| Glass bottle | |
| MFCD00041790 | |
| 26, 420 | |
| adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
| Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in acids and alkalies,slightly soluble in ethanol,insoluble in chloroform and ether,25g/l in boiling water | |
| GFFGJBXGBJISGV-UHFFFAOYSA-N | |
| 7H-purin-6-amine | |
| 190 | |
| 135.13 | |
| Liquid or Solid |
| 73-24-5 | |
| 0.74 to 0.78 (A250/A260), 0.36 to 0.4 (A280/A260) | |
| 360°C to 365°C | |
| 0.5% max. (105°C, 5 hrs) | |
| 98.5% min. (HPLC) | |
| C5H5N5 | |
| 25 g | |
| 15, 140 | |
| 220°C | |
| 98.0 to 102.0% (lambda max. 262nm, pH 1), molar absorptivity 13100 +/- 2%, on dry basis | |
| NC1=C2NC=NC2=NC=N1 | |
| 135.13 | |
| CHEBI:16708 | |
| 99% |
Chemical Identifiers
| 73-24-5 | |
| 135.13 | |
| GFFGJBXGBJISGV-UHFFFAOYSA-N | |
| 190 | |
| 7H-purin-6-amine |
| C5H5N5 | |
| MFCD00041790 | |
| adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
| CHEBI:16708 | |
| NC1=C2NC=NC2=NC=N1 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 200-796-1
RUO – Research Use Only