missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Adenine, ≥98%, Calbiochem™,
Supplier: MilliporeSigma™ 115225GM
Specifications
| 73-24-5 | |
| Glass Bottle | |
| MFCD00041790 | |
| adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
| NC1=C2NC=NC2=NC=N1 | |
| 135.13 | |
| CHEBI:16708 | |
| Solid |
| White | |
| C5H5N5 | |
| 25 g | |
| GFFGJBXGBJISGV-UHFFFAOYSA-N | |
| 7H-purin-6-amine | |
| 190 | |
| >98% by Assay; ≥99% by HPLC |
Chemical Identifiers
| 73-24-5 | |
| 135.13 | |
| GFFGJBXGBJISGV-UHFFFAOYSA-N | |
| 190 | |
| 7H-purin-6-amine |
| C5H5N5 | |
| MFCD00041790 | |
| adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine | |
| CHEBI:16708 | |
| NC1=C2NC=NC2=NC=N1 |