missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Adenine, ≥98%, Calbiochem™,

Catalog No. 115210GM
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
115210GM 10 g Glass Bottle
115225GM 25 g Glass Bottle
1152100GM 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 115210GM Supplier MilliporeSigma™ Supplier No. 115210GM

Chemical Identifiers

CAS 73-24-5
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00041790
InChI Key GFFGJBXGBJISGV-UHFFFAOYSA-N
Synonym adenine, 1h-purin-6-amine, 6-aminopurine, 9h-purin-6-amine, vitamin b4, adenin, adeninimine, leuco-4, 6-amino-1h-purine, 6-amino-3h-purine
PubChem CID 190
ChEBI CHEBI:16708
IUPAC Name 7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC=N1

Specifications

Color White
Packaging Glass Bottle
Quantity 10 g
Percent Purity >98% by Assay; ≥99% by HPLC
Physical Form Solid