Learn More
Thermo Scientific™ Adapters for TX-150 Swinging Bucket Rotors
Assorted tube adapters
Supplier: Thermo Scientific™ 75003504
Description
Assorted tube adapters
Specifications
| Rotor Adapter | |
| Polypropylene | |
| 17 mm | |
| TX-150 rotor (75005701) | |
| 4 |
| 13 mL Urine Tube | |
| 4 x 13 mL | |
| 110 mm | |
| Adapters for 13 mL urine tubes used in TX-150 rotor | |
| 4 |