Learn More
Thermo Scientific™ Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Supplier:  Thermo Scientific™ 75007304
 View more versions of this product
Catalog No. 75007304
Only null left
Description
Description
An assortment of adapters for outstanding flexibility
Specifications
Specifications
Rotor Adapter
4 x 750 mL
133 mm
750 mL Bio Bottle
98 mm
750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
Product Suggestions
Product Suggestions