missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adapters for Round Single Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors

Catalog No. 75003858
Change view
Click to view available options
Capacity:
6/8 x 450 mL Single or Double Blood Bag
6/8 x 500 mL Triple or Quad Blood Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003858 6/8 x 450 mL Single or Double Blood Bag
75003857 6/8 x 500 mL Triple or Quad Blood Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75003858 Supplier Thermo Scientific™ Supplier No. 75003858

Select adapters for 450-500mL blood bags in round single blood bag buckets for Thermo Scientific™ HAEMAFlex™ 6/8 rotors.

Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.

Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor or, utilizing the same bucket system, upgrade to the HAEMAFlex 8 swinging bucket rotor.

Select adapters for 450–500mL Triple or Quad blood bags or 450mL Single or Double blood bags.

Specifications

Type Blood Bag Adapter
Capacity 6/8 x 450 mL Single or Double Blood Bag
For Use With (Equipment) Single blood bag bucket with filter pack
Description 450mL Single or Double Blood Bag Adapter (set of 2)