Learn More
Chemglass Life Sciences Glass Adapter Only, 24/40, Glass DFS Item
Supplier:  Chemglass Inc CG131810
Glass Adapter Only, 24/40, Glass. Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator. Adapters have a top outer joint of the listed size and a 20-400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.
Catalog No. CG131810
Only null left
Explore available promotions