Learn More
Synthware™ Enlarging Adapters
With standard taper joints at top and bottom
Supplier:  Synthware™ A253424
 View more versions of this product
Catalog No. 31500518
Only null left
Explore available promotions
Description
Description
Synthware enlarging adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is smaller in size than the top outer joint.
Specifications
Specifications
Organic Synthesis
Enlarging Adapter
Top: 34/45, Bottom: 24/40
Borosilicate Glass (3.3)
1
Product Suggestions
Product Suggestions