Learn More
Synthware™ Enlarging Adapters
With standard taper joints at top and bottom
Supplier: Synthware™ A252414
Description
Synthware enlarging adapters are used for connecting dissimilar standard taper joint sizes. The lower inner joint is smaller in size than the top outer joint.Specifications
| Organic Synthesis | |
| Enlarging Adapter | |
| Top: 24/40, Bottom: 14/20 |
| Borosilicate Glass (3.3) | |
| 1 |