Learn More
Chemglass Life Sciences Adapter, Bushing, 34/45 Lower Inner, 24/40 Top Outer DFS Item
Supplier:  Chemglass Inc CG101612
Adapter, Bushing, 34/45 Lower Inner, 24/40 Top Outer. Bushing style adapter having a lower standard taper inner joint and outer joint ground in the bore of the adapter. Adapters having a 10/18 or 10/30 outer member in the bore are used to insert thermometers into apparatus.
Catalog No. CG101612
Only null left
Explore available promotions