Learn More
Synthware™ Adapter, Joint to Inner Joint
Adapter, No. 15 O-ring joint to 14/20, 19/22 or 24/40 inner joint
$87.21 - $92.89
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Standard Taper Joint No. | Price | Quantity & Availability | |||||
31501811
![]() |
Synthware™
M131514 |
14/20 |
Each for $87.21
|
|
|||||
31501812
![]() |
Synthware™
M131519 |
19/22 |
Each for $90.71
|
|
|||||
31501813
![]() |
Synthware™
M131524 |
24/40 |
Each for $92.89
|
|
|||||
Specifications
| Organic Synthesis | |
| Borosilicate Glass | |
| 1 Ea. |