missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter
Adapter for rectangular bucket in Rotor A-4-62.
Supplier: Eppendorf™ 022638424
Description
- For 250 mL rectangular bucket in Rotor A-4-62
- 2 pcs.
Chemical Identifiers
| Adapter | |
| Rotor A-4-62 |
Specifications
| Adapter | |
| 1 Tube | |
| Rotor A-4-62 | |
| 80 to 120 mL |