missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter

Catalog No. 05414141
Change view
Click to view available options
Capacity:
1 Tube
2 Tubes
4 Tubes
Tube Size:
18 to 30 mL
50 to 75 mL
80 to 120 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Tube Size
05414141 2 Tubes 50 to 75 mL
05414140 4 Tubes 18 to 30 mL
05414142 1 Tube 80 to 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05414141 Supplier Eppendorf™ Supplier No. 022638408
Only null left

Adapter for rectangular bucket in Rotor A-4-62.

  • For 250 mL rectangular bucket in Rotor A-4-62
  • 2 pcs.

Specifications

Type Adapter
Tube Type Round Bottom
Capacity 2 Tubes
For Use With (Equipment) Rotor A-4-62
Tube Size 50 to 75 mL