missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter

Catalog No. 05414167
Change view
Click to view available options
For Use With (Equipment):
Rotor F-45-18-17-Cryo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05414167 Rotor F-45-18-17-Cryo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05414167 Supplier Eppendorf™ Supplier No. 022639528
Only null left

Adapter for Rotor F-45-18-17-Cryo.

  • For 1 cryo tube (max. diameter 13 mm) or sealable centrifugation tubes (diameter 12.2 mm)
  • Max. length 50 mm
  • For Rotor F-45-18-17-Cryo
  • 6 pcs.

Specifications

Type Adapter
Tube Type Cryotube
Max. Tube Diameter 13 mm
Max. Tube Length 50 mm
For Use With (Equipment) Rotor F-45-18-17-Cryo
Vessels per Adapter 1