Learn More
Eppendorf™ Adapter
Description
- Adapter for:
- 20 reaction vessels 1.5/2.0 mL
- 500 mL rectangular bucket in Rotor A-4-81 and Rotor S-4x500
- 2 pcs
Specifications
Specifications
| Type | Adapter |
| For Use With (Equipment) | Rotor A-4-81 and Rotor S-4x500 |
| Vessels per Adapter | 20 Reaction Vessels |
| Tube Size | 1.5/2.0 mL |