Learn More
Chemglass Life Sciences Adapter, inlet, inner joint, straight. Joint Size: 45/50, Overall Length(mm): 110
Supplier: Chemglass Inc CG1012C05
Adapter, inlet, inner joint, straight. Joint Size: 45/50, Overall Length(mm): 110. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.