missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter for 250 mL Rectangular Bucket in Rotor FA - 6 x 250
Adapts a wide range of centrifuge tubes for use in Rotor FA-6x250.
$750.54 - $750.54
Specifications
| Description | For 250 mL rectangular bucket in Rotor FA-6x250 |
|---|---|
| No. per Pack | Set of 2 pieces |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity | |||||
|
05-413-909
|
Eppendorf™
5920767003 |
3 x 30 mL Oak Ridge Round Centrifuge Tubes |
Each for $750.54
|
|
|||||
Description
- Adapts a wide range of centrifuge tubes to fit in Rotor FA - 6 x 250
- Set of 2 pieces
Specifications
| For 250 mL rectangular bucket in Rotor FA-6x250 | |
| Set of 2 pieces |