missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapter with 10-32 UNF Thread x 3/32 in. Barb - Polypropylene

Catalog No. 01000002
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01000002 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000002

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Material Non-Animal Derived Polypropylene
For Use With (Application) Threaded Tube Connector for Medical, Pharmaceutical, Industrial applications.
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Thread Adapter
Quantity 10 Pk.