missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carboxylic acid, 99%

CAS: 828-51-3 | C11H16O2 | 180.247 g/mol

Supplier:  Thermo Scientific Chemicals A1295922

 View more versions of this product

Catalog No. AAA1295922

Only null left
Explore available promotions

Description

Description

It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a stabilizer in the synthesis of monodisperse, highly crystalline CoPt3 nanoparticles1 and porous platinum nanoparticles. It is a potent and reversible CerK inhibitor.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in ethanol, chloroform, and dichloromethane.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Adamantane-1-carboxylic acid
173°C to 176°C
MFCD00074720
1910637
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in ethanol,chloroform,and dichloromethane.
C1C2CC3CC1CC(C2)(C3)C(=O)O
180.247
180.25
828-51-3
C11H16O2
100 g
1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
JIMXXGFJRDUSRO-UHFFFAOYSA-N
adamantane-1-carboxylic acid
13235
99%

Chemical Identifiers

828-51-3
180.247
JIMXXGFJRDUSRO-UHFFFAOYSA-N
13235
C1C2CC3CC1CC(C2)(C3)C(=O)O
C11H16O2
MFCD00074720
1-adamantanecarboxylic acid, 1-carboxyadamantane, adamantanecarboxylic acid, adamantoic acid, 1-adamantoic acid, 3-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, tricyclo 3.3.1.1'3,7 decane-1-carboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3r,5s,7s-adamantane-1-carboxylic acid
adamantane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-584-6

RTECSNumber : AU4452200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only